Anti-GEMIN6 Rabbit Polyclonal Antibody
AVIVARP62117_P050
- Antibody type:Primary
- Antigen name:Gem (nuclear organelle) associated protein 6
- Antigen symbol:GEMIN6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:79833
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: GEMIN6 (gem (nuclear organelle) associated protein 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: