Anti-TIPRL Rabbit Polyclonal Antibody
Catalog # AVIVARP62048_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:TOR signaling pathway regulator
- Antigen symbol:TIPRL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:261726
- Antigen synonyms:TIP|TIP41
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TIPRL (TOR signaling pathway regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: