Anti-CES3 Rabbit Polyclonal Antibody
AVIVARP62006_P050
- Antibody type:Primary
- Antigen name:Carboxylesterase 3
- Antigen symbol:CES3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:23491
- Antigen synonyms:ES31
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: CES3 (carboxylesterase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human