Anti-CISH Rabbit Polyclonal Antibody
Catalog # AVIVARP52798_P050
Dodavatel: Aviva Systems Biology
Specifikace
- Antibody type:Primary
- Antigen name:cytokine inducible SH2-containing protein
- Antigen symbol:CISH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:12700
- Antigen synonyms:CIS1|SOCS|Cis|F23|CIS-1|F17
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifikace
O tomto produktu
Type: Primary
Antigen: CISH (cytokine inducible SH2-containing protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: