Anti-KCNT1 Rabbit Polyclonal Antibody
AVIVARP49576_P050
- Antibody type:Primary
- Antigen name:potassium channel, subfamily T, member 1
- Antigen symbol:KCNT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:227632
- Antigen synonyms:Slo2.2|Slack|C030030G16Rik|slo2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: KCNT1 (potassium channel, subfamily T, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: