Anti-LRCH4 Rabbit Polyclonal Antibody
Katalogové číslo AVIVARP45267_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:leucine-rich repeats and calponin homology (CH) domain containing 4
- Antigen symbol:LRCH4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:4034
- Antigen synonyms:LRRN1|LRRN4|PP14183|LRN
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: LRCH4 (leucine-rich repeats and calponin homology (CH) domain containing 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: