Anti-OR2W5 Rabbit Polyclonal Antibody
Katalogové číslo AVIVARP44489_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:olfactory receptor, family 2, subfamily W, member 5 (gene/pseudogene)
- Antigen symbol:OR2W5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:441932
- Antigen synonyms:OR2W5P|OST722
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: OR2W5 (olfactory receptor, family 2, subfamily W, member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human