Anti-TMEM200B Rabbit Polyclonal Antibody
Catalog # AVIVARP44447_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:transmembrane protein 200B
- Antigen symbol:TMEM200B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:399474
- Antigen synonyms:TTMB
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TMEM200B (transmembrane protein 200B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: