Anti-CES1 Rabbit Polyclonal Antibody
Catalog # AVIVARP41877_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Carboxylesterase 1
- Antigen symbol:CES1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Bovine,Guinea pig,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:1066
- Antigen synonyms:CES2|CEH|HMSE|SES1|REH|hCE-1|CE-1|ACAT|HMSE1|TGH|PCE-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CES1 (carboxylesterase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Rat