Anti-KDM2B Rabbit Polyclonal Antibody
AVIVARP39706_P050
- Antibody type:Primary
- Antigen name:lysine (K)-specific demethylase 2B
- Antigen symbol:KDM2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:84678
- Antigen synonyms:PCCX2|JHDM1B|CXXC2|Fbl10|FBXL10
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: KDM2B (lysine (K)-specific demethylase 2B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: