Anti-LIMA1 Rabbit Polyclonal Antibody
AVIVARP39256_P050
- Antibody type:Primary
- Antigen name:LIM domain and actin binding 1
- Antigen symbol:LIMA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:51474
- Antigen synonyms:SREBP3|EPLIN
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: LIMA1 (LIM domain and actin binding 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: