Anti-GTF2IRD1 Rabbit Polyclonal Antibody
Katalogové číslo AVIVARP39254_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:GTF2I repeat domain containing 1
- Antigen symbol:GTF2IRD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:9569
- Antigen synonyms:RBAP2|hMusTRD1alpha1|WBSCR11|WBS|CREAM1|GTF3|MUSTRD1|BEN|WBSCR12
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: GTF2IRD1 (GTF2I repeat domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: