Anti-HEY2 Rabbit Polyclonal Antibody
Catalog # AVIVARP38979_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Hes-related family bHLH transcription factor with YRPW motif 2
- Antigen symbol:HEY2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:23493
- Antigen synonyms:GRL|bHLHb32|GRIDLOCK|HESR2|HRT2|HERP1|CHF1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: HEY2 (hes-related family bHLH transcription factor with YRPW motif 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: