Anti-RYBP Rabbit Polyclonal Antibody
AVIVARP38973_P050
- Antibody type:Primary
- Antigen name:RING1 and YY1 binding protein
- Antigen symbol:RYBP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:23429
- Antigen synonyms:AAP1|DEDAF|YEAF1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: RYBP (RING1 and YY1 binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: