Anti-CIZ1 Rabbit Polyclonal Antibody
AVIVARP38956_P050
- Antibody type:Primary
- Antigen name:CDKN1A interacting zinc finger protein 1
- Antigen symbol:CIZ1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:25792
- Antigen synonyms:NP94|ZNF356|LSFR1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: CIZ1 (CDKN1A interacting zinc finger protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: