Order Entry
Czech Republic
ContactUsLinkComponent
Anti-POU4F1 Rabbit Polyclonal Antibody
Anti-POU4F1 Rabbit Polyclonal Antibody
  AVIVARP38763_T100
Anti-POU4F1 Rabbit Polyclonal Antibody
  AVIVARP38763_T100

 

  • Antibody type:
    Primary
  • Antigen name:
    POU class 4 homeobox 1
  • Antigen symbol:
    POU4F1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Gene ID:
    5457
  • Antigen synonyms:
    BRN3A|RDC-1|Oct-T1|brn-3A
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: POU4F1 (POU class 4 homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: