Anti-MED7 Rabbit Polyclonal Antibody
Catalog # AVIVARP38446_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Mediator Complex Subunit 7
- Antigen symbol:MED7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:9443
- Antigen synonyms:CRSP9|CRSP33|ARC34
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: MED7 (mediator complex subunit 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: