Anti-MED27 Rabbit Polyclonal Antibody
Catalog # AVIVARP38444_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Mediator Complex Subunit 27
- Antigen symbol:MED27
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:9442
- Antigen synonyms:CRSP8|CRSP34|TRAP37|CRAP34
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: MED27 (mediator complex subunit 27)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: