Anti-PITX2 Rabbit Polyclonal Antibody
AVIVARP37981_P050
- Antibody type:Primary
- Antigen name:Paired-like homeodomain 2
- Antigen symbol:PITX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:5308
- Antigen synonyms:IDG2|RIEG1|IHG2|IGDS2|RIEG|RS|Brx1|IRID2|IGDS|ARP1|RGS|PTX2|Otlx2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: PITX2 (paired-like homeodomain 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: