Anti-AIP Rabbit Polyclonal Antibody
Katalogové číslo AVIVARP37734_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Aryl hydrocarbon receptor interacting protein
- Antigen symbol:AIP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:9049
- Antigen synonyms:FKBP37|FKBP16|XAP2|XAP-2|ARA9|SMTPHN
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: AIP (aryl hydrocarbon receptor interacting protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: