Anti-G3BP1 Rabbit Polyclonal Antibody
Catalog # AVIVARP37713_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:GTPase activating protein (SH3 domain) binding protein 1
- Antigen symbol:G3BP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:10146
- Antigen synonyms:G3BP|HDH-VIII
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: G3BP1 (GTPase activating protein (SH3 domain) binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: