Anti-KCNQ5 Rabbit Polyclonal Antibody
Catalog # AVIVARP37700_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:potassium channel, voltage gated KQT-like subfamily Q, member 5
- Antigen symbol:KCNQ5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:56479
- Antigen synonyms:Kv7.5
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: KCNQ5 (potassium voltage-gated channel, KQT-like subfamily, member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: