Anti-TJP1 Rabbit Polyclonal Antibody
Catalog # AVIVARP36606_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Tight Junction Protein 1
- Antigen symbol:TJP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:292994
- Antigen synonyms:ZO-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TJP1 (tight junction protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: