Anti-ANXA13 Rabbit Polyclonal Antibody
Catalog # AVIVARP36590_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Annexin A13
- Antigen symbol:ANXA13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Pig,Human,Bovine,Guinea pig,Rat,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:312
- Antigen synonyms:ISA|ANX13
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ANXA13 (annexin A13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Guinea pig, Human, Mouse, Pig, Rat