Anti-ANXA6 Rabbit Polyclonal Antibody
Katalogové číslo AVIVARP36583_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:annexin A6
- Antigen symbol:ANXA6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:309
- Antigen synonyms:CBP68|ANX6
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ANXA6 (annexin A6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: