Anti-KCNJ4 Rabbit Polyclonal Antibody
AVIVARP35167_P050
- Antibody type:Primary
- Antigen name:potassium channel, inwardly rectifying subfamily J, member 4
- Antigen symbol:KCNJ4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:3761
- Antigen synonyms:Kir2.3|IRK-3|IRK3|HIR|HRK1|HIRK2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: KCNJ4 (potassium inwardly-rectifying channel, subfamily J, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: