Anti-KCNQ2 Rabbit Polyclonal Antibody
Catalog # AVIVARP35140_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:potassium voltage-gated channel, subfamily Q, member 2
- Antigen symbol:KCNQ2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:16536
- Antigen synonyms:HNSPC|KQT2|Nmf134
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: KCNQ2 (potassium voltage-gated channel, KQT-like subfamily, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: