Anti-CACNB2 Rabbit Polyclonal Antibody
AVIVARP34956_P050
- Antibody type:Primary
- Antigen name:calcium channel, voltage-dependent, beta 2 subunit
- Antigen symbol:CACNB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:783
- Antigen synonyms:CACNLB2|MYSB|CAVB2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: CACNB2 (calcium channel, voltage-dependent, beta 2 subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: