Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-RNF135 Rabbit Polyclonal Antibody
  AVIVARP34885_P050
Anti-RNF135 Rabbit Polyclonal Antibody
  AVIVARP34885_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    Ring Finger Protein 135
  • Antigen symbol:
    RNF135
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    middle
  • Cross adsorption:
    none
  • Gene ID:
    84282
  • Antigen synonyms:
    L13|Riplet|MMFD|REUL
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: RNF135 (ring finger protein 135)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human