Anti-CCNL1 Rabbit Polyclonal Antibody
Catalog # AVIVARP33174_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:cyclin L1
- Antigen symbol:CCNL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:57018
- Antigen synonyms:ANIA6A|PRO1073|BM-001|ania-6a
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CCNL1 (cyclin L1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: