Anti-MYCN Rabbit Polyclonal Antibody
Catalog # AVIVARP32030_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:v-myc avian myelocytomatosis viral oncogene neuroblastoma derived homolog
- Antigen symbol:MYCN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:4613
- Antigen synonyms:ODED|NMYC|N-myc|MODED|bHLHe37
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: MYCN (v-myc avian myelocytomatosis viral oncogene neuroblastoma derived homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: