Anti-CD81 Rabbit Polyclonal Antibody
Catalog # BOSSBS-6934R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:CD81 molecule
- Antigen symbol:CD81
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:CD81 antigen|S5.7|TSPAN28|26 kDa cell surface protein TAPA-1|CVID6|Tetraspanin-28|TAPA1|Target of Antiproliferative 1|TSPAN-28
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CD81
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: