Anti-LTB4R2 Rabbit Polyclonal Antibody
Catalog # BOSSBS-2655R
Dodavatel: Bioss
Specifikace
- Antibody type:Primary
- Antigen name:Leukotriene B4 Receptor 2
- Antigen symbol:LTB4R2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:NOP9|BLTR2|KPG_004|LTB4-R 2|JULF2|LTB4-R2|BLT2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifikace
O tomto produktu
Type: Primary
Antigen: LTB4R2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat