Anti-TNFSF14 Rabbit Polyclonal Antibody
BOSSBS-2462R
: Bioss
- Antibody type:Primary
- Antigen name:Tumor Necrosis Factor Superfamily Member 14
- Antigen symbol:TNFSF14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:CD258|LTg|LIGHT|HVEML
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Type: Primary
Antigen: TNFSF14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat