Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-CK1+5+10+14 Rabbit polyclonal antibody
Catalog # BOSSBS-1494R
Supplier:  Bioss
Anti-CK1+5+10+14 Rabbit polyclonal antibody
Catalog # BOSSBS-1494R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CK1+5+10+14
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Pig,
    Human,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: CK1+5+10+14
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Human, Mouse, Pig, Rat