Order Entry
Czech Republic
ContactUsLinkComponent
Anti-HSP90AB1 Rabbit Polyclonal Antibody
Anti-HSP90AB1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-0194R
Supplier:  Bioss
undefined
Anti-HSP90AB1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-0194R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Heat Shock Protein 90kDa Alpha (Cytosolic), Class B Member 1
  • Antigen symbol:
    HSP90AB1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Antigen synonyms:
    HSP84|HSPCB|HSPC2|Heat shock protein HSP 90-beta|HSP 90|HSP90B|Heat shock 84 kDa|HSP 84|HSP90AB1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: HSP90AB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat