ContactUsLinkComponent
Filter holder assembly, All-Glass
Filter holder assembly, All-Glass
Catalog # GLAS258.202.01
Supplier:  GLASSCO
undefined
Filter holder assembly, All-Glass
Catalog # GLAS258.202.01
Supplier:  GLASSCO
Specifications
  • Description:
    Glass filter assembly with 300 ml funnel, 1000 ml flask, fritted base, cap and clamp, 47 mm
  • Pk:
    1
Specifications
About this item
Vacuum filter for aqueous, organic and corrosive liquids for particulate contamination analysis. Also recommended for HPLC solvent filtration.