Peqlab, PCR Plates, 96-Well, Semi-Skirted
Supplier: VWR Collection
Certificates
About this item
PP. Compatible with all major thermal cyclers.
- Greater rigidity than non-skirted plates and provides space for barcoding
- Plates with segmented design can be cut into 24 and 48 well sections
- Ultra-thin for effective heat transfer and minimum condensation
- Cut away corner for easy orientation
- Certified free from RNase, DNase and human DNA
Specifications
- Catalog No:
- 732-3201
- 732-3227
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)