Capillary columns, Ultra 2
: Agilent
Columns equivalent to HP-5 with tighter specifications for retention index and capacity factors.
- Non polar
- Bonded and cross-linked
- Solvent rinsable
(5%-Phenyl)-methylpolysiloxane
Frequently Bought Together
Columns equivalent to HP-5 with tighter specifications for retention index and capacity factors.
(5%-Phenyl)-methylpolysiloxane