ContactUsLinkComponent
Floating racks for cyrotubes, Bel-Art ProCulture, SP Bel-Art
Floating racks for cyrotubes, Bel-Art ProCulture, SP Bel-Art
Catalog # BELAF188760020
undefined
Floating racks for cyrotubes, Bel-Art ProCulture, SP Bel-Art
Catalog # BELAF188760020
Specifications
  • Description:
    Cyrotube floating racks, 20 places, PP, Ø 95 mm
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
This 20 place polypropylene round floating rack fits most cryotubes and holds them tightly in a circular pattern formed by 12 outer and 8 inner openings.
  • Floats in water; can also be used for quickly thawing frozen cells in a 37 ºC water bath
  • Easily remove tubes by holding the center handle and pushing the rack down against the bench until the legs touch
  • Dimensions: 6,4 mm (¹/₄") thick; 95 mm (3³/₄") in Ø; 19 mm (³/₄") long legs
  • Autoclavable
Excellent for holding cryotubes during work or incubation.