ContactUsLinkComponent
Cleaning cloths, Praktitex® Pro
Cleaning cloths, Praktitex® Pro
Catalog # ZVGA10294-01
undefined
Cleaning cloths, Praktitex® Pro
Catalog # ZVGA10294-01
Specifications
  • Packed:
    10 wipes/pack
  • Colour:
    Pink
  • L×W:
    400×400 mm
  • Environmentally Preferable:
  • Pk:
    10
Specifications
About this item
This microfibre cloth (80% polyester, 20% polyamide) has an open structure and is therefore ideal for cleaning heavily fouled surfaces.
  • Extremely fine fibres penetrate deep into the surface and thus effectively loosen adhering and grease-containing fouling
  • Washable and therefore reusable
Weight: 300 g/m²