Accessories for vacuum filtration devices GV 050 series, Whatman™
Supplier: Whatman products (Cytiva)
Certificates
Accessories for vacuum filtration devices GV 050 series, Whatman™
Supplier: Whatman products (Cytiva)
About this item
Frequently Bought Together
Quantitative filter papers, ashless grades, grade 589/1 and 589/1 ½, Whatman™
Supplier: Whatman products (Cytiva)
VWR® Weighing Paper, Grade 531
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Methanol, anhydrous (max. 0.003% H₂O) analytical reagent
Supplier: Merck
VWR®, Syringe Filters
Supplier: VWR Collection
pH electrodes, Sentix® IDS
Supplier: WTW
Vacuum filtering flasks, Büchner, with glass hose connection
Supplier: DWK Life Sciences
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Single use syringes, 2-piece, Injekt® Solo
Supplier: B. Braun
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Diethyl ether ≥99.8% contains inhibitor, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Accessories for vacuum filtration devices GV 050 series, Whatman™
Supplier: Whatman products (Cytiva)