ContactUsLinkComponent
Visual colour kits, phosphorus, PO-19A
Visual colour kits, phosphorus, PO-19A
Catalog # HACH224801
Supplier:  Hach
CAS Number:  
undefined
Visual colour kits, phosphorus, PO-19A
Catalog # HACH224801
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Phosphorus, orthophosphate test kit, model PO-19A
  • Range:
    0 - 0,8 mg/l PO₄
    0 - 4 mg/l PO₄
    0 - 40 mg/l PO₄
Specifications
About this item
Simple, economical rapid colour disc method with an easy to carry test kit.
Delivery: Kit includes two reagents, colour disc, comparator box, two viewing tubes, viewing adaptor, two bottles, dropper tube, funnel, filter paper, deionized water, instructions and carrying case.