ContactUsLinkComponent
Multi parameter test kit
Multi parameter test kit
Catalog # HACH223002
Supplier:  Hach
CAS Number:  
undefined
Multi parameter test kit
Catalog # HACH223002
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Multi parameter test kit, CN-39WR
Specifications
About this item
This multi-test kit combines popular water treatment tests into one compact kit which can be used to determine chlorine (0 to 3,4 mg/L), hardness (1 to 20 gpg), iron (0 to 4 mg/L) and pH (4 to 10) in the water sample.
  • Includes frequently measured water quality parameters
  • Uses simple, economical methods
Hardeness testing: gpg= grains per gallon
Delivery: Includes six reagents, bottles, colour discs, comparator box, viewing tubes, measuring tube, instructions and carrying case.