
Lithium chloride, peqGOLD OptiPure
Catalog # 732-3315
Supplier: VWR Collection
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:42.39 g/mol
EINECS:231-212-3
:GHS07
Storage Temperature:Ambient
CAS Number:7447-41-8
Formula:LiCl
MDL Number:MFCD00011078