ContactUsLinkComponent
Visual comparator colour kits, phenols
Visual comparator colour kits, phenols
  HACH2483600
 :  Hach
 :  
undefined
Visual comparator colour kits, phenols
  HACH2483600
 :  Hach
 :  
  • Chemical method:
    4-Aminoantipyrine
  • Pk:
    1 KIT
  • Description:
    Phenols test kit, model PL-1
  • Range:
    0 - 4 mg/l
Simple, economical, rapid visual colour determination.
 : Kit includes four reagents, two tubes, two color discs, color comparator box, instruction sheet, and carrying case.