ContactUsLinkComponent
Chromium test kits, models CH-8 and CH-12, comparator method
Chromium test kits, models CH-8 and CH-12, comparator method
Catalog # HACH183400
Supplier:  Hach
CAS Number:  
undefined
Chromium test kits, models CH-8 and CH-12, comparator method
Catalog # HACH183400
Supplier:  Hach
CAS Number:  
Specifications
  • Chemical method:
    Diphenylcarbazide hypobromite oxidation
  • Pk:
    1 KIT
  • Description:
    Chromium (VI) test kit, model CH-8, colour disc
  • Range:
    0 - 1,3 mg/l
Specifications
About this item
These kits are used to determine chromium or chromium(VI) using simple, economical colour disc methods.
Delivery: CH-12 kit includes four reagents, clippers, two colour discs, comparator box, two viewing tubes, bottle, flask, dropper tube, stove and cover, test tube clamp, instructions and carrying case. CH-8 kit includes one reagent, two colour discs, comparator box, two viewing tubes, vial, instructions and carrying case.