ContactUsLinkComponent
Aluminium reagent set for Al determination by photometric ECR method
Aluminium reagent set for Al determination by photometric ECR method
Catalog # HACH2443400
Supplier:  Hach
CAS Number:  
undefined
Aluminium reagent set for Al determination by photometric ECR method
Catalog # HACH2443400
Supplier:  Hach
CAS Number:  
Specifications
  • Chemical method:
    Eriochrome cyanine R
  • Pk:
    1 SET
  • Description:
    Aluminium reagent set, 100 tests
  • For unit:
    DR2000, DR3000
  • Range:
    0,002 - 0,220 mg/l
Specifications
About this item
Kit used for the determination of aluminium by Eriochrome Cyanine R (ECR) method and has sample size of 50 ml.
Delivery: Set contains ECR reagent powder pillows, hexamethylenetetramine buffer reagent powder pillows and ECR masking reagent solution for approximately 100 tests.