Order Entry
Czech Republic
ContactUsLinkComponent
BCIP/NBT alkaline phosphatase substrate
BCIP/NBT alkaline phosphatase substrate
Catalog # ROCKNBT-100
CAS Number:  
BCIP/NBT alkaline phosphatase substrate
Catalog # ROCKNBT-100
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

Specification Test Results

  • pH:
    9.28 +/- 0.1
  • QC Raw Material:
    PASS
  • Absorbance check of final product:
    PASS
  • Performance Data per Immunoblot:
    PASS

Specification Test Results

About this item

Synonyms: BCIP (5-bromo-4-chloro-3-indolylphosphate toluidine salt), NBT (nitro-blue tetrazolium chloride)
 :
Storage Temperature:Refrigerator