Order Entry
Czech Republic
ContactUsLinkComponent
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]), liquid ELISA peroxidase substrate, ready-to-use
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]), liquid ELISA peroxidase substrate, ready-to-use
Catalog # ROCKABTS-100
CAS Number:  
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]), liquid ELISA peroxidase substrate, ready-to-use
Catalog # ROCKABTS-100
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :548.68 g/mol
Storage Temperature:Refrigerator
CAS Number:30931-67-0
Formula:C₁₈H₂₄N₆O₆S₄
MDL Number:MFCD00010404